Learn More
Thermo Scientific™ HRP-Conjugated Streptavidin
Streptavidin from Streptomyces avidinii conjugated to highly purified horseradish peroxidase (HRP)
Brand: Thermo Scientific™ N100
83.33 GBP valid until 2024-06-29
Use promo code "21620" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.68040kg
Description
- Validated for use with Matched Antibody Pairs and MiniKits
- Pre-defined working ranges save time and facilitate assay optimization
Specifications
1.25 mg/mL | |
Store between 2°C and 8°C. | |
Streptavidin Conjugate | |
HRP (Horseradish Peroxidase) |
HRP | |
Liquid | |
Biotin |
Safety and Handling
- HRP-Conjugated Streptavidin
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 5-chloro-2-methyl4-isothiazolin-3-one [EC no. 247-500-7] and2-methyl-2Hisothiazol-3-one
For Research Use Only. Not for use in diagnostic procedures.