missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ No-Weigh™ Sulfo-SMCC
Format preserves reagent activity and prevents cross-contamination between uses
Brand: Thermo Scientific™ 22622
This item is not returnable.
View return policy
Description
- Reagents are dispensed and dried in individual microtubes
- Heat sealed with a foil that can be easily punctured with a pipet tip
- Losses are minimized since the reagent can be reconstituted just before it is needed for the labeling reaction
- Water soluble, so crosslinking can be done in physiologic solutions
- Amine reactive Sulfo-NHS ester crosslinks rapidly with primary amine-containing molecule Sulfhydryl-reactive maleimide reacts with cysteine residues to yield specific conjugates High purity, crystalline Sulfo-SMCC can be used to create high-purity maleimide-activated derivatives Cyclohexane bridge confers added stability to the maleimide group making Sulfo-SMCC an excellent crosslinking agent for maleimide activation of proteins
- Maleimide groups are stable for 64 hours in 0.1M sodium phosphate buffer, pH 7 at 4°C
- CAS: 92921-24-9
- Spacer Arm 8.3Å
Specifications
OSHA, 29 CFR 1910.1200 | |
Sulfo-SMCC 98 to 100% | |
37mL of typical 1X (1mM) reaction solution (to tal) or 8 separate uses | |
2 mg |
Sulfosuccinimidyl 4- (N-maleimidomethyl) cyclohexane-1-carboxylate, Solid | |
436.37 | |
-20°C | |
Sulfo-SMCC |