Learn More
Thermo Scientific™ Poly-HRP Dilution Buffer
Specifically designed to optimize performance of Poly-HRP Streptavidin
Brand: Thermo Scientific™ N500
Code : PN
Additional Details : CAS Number : 626-67-5 Weight : 0.05000kg Transport : UN number : 2399 Chem class : 3 Pack group : II
Description
- 1% biotin-free casein in a PBS buffer solution
- Ready-to-use
Specifications
Dilution Buffer | |
Shipped on cold packs. Store at 2°C to 8°C immediately upon receipt. | |
Liquid |
Diluent | |
100 mL |
For Research Use Only. Not for use in diagnostic procedures.