missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Poly-HRP Streptavidin
An advanced polymer of HRP conjugated to streptavidin with a high HRP:streptavidin ratio
Brand: Thermo Scientific™ N200
261.65 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Produces an enhanced signal
- May increase ELISA sensitivity
- Validated for use with some Thermo Scientific Pierce Matched Antibody Pairs and MiniKits
Specifications
1.0 mg/mL | |
0.25 mL | |
Liquid | |
Biotin |
HRP | |
Do not store below -20°C. Avoid multiple freeze-thaw cycles. | |
Streptavidin Conjugate | |
HRP (Horseradish Peroxidase) |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Poly-HRP Streptavidin
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : glycerol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction