Learn More
Thermo Scientific™ TMB Substrate Solution
A ready-to-use substrate system containing 3.3', 5.5'-tetramethylbenzidine (TMB)
Brand: Thermo Scientific™ N301
109.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.
Code : PN
Additional Details : Weight : 0.10000kg
Description
- Ready to use, no preparation required
- Validated for use with Thermo Scientific Pierce Matched Antibody Pairs and MiniKits
- Size: 100 mL (sufficient for about ten 96-well microplates)
Specifications
Chemical Substrate | |
Colorimetric | |
HRP (Horseradish Peroxidase) Substrate | |
100 mL |
Store between 2°C and 8°C. | |
Pierce™ | |
TMB Substrate |
For Research Use Only. Not for use in diagnostic procedures.