Learn More
Thermo Scientific™ QCC (Quick Column Change) for Agilent Instruments
This Quick Column Change accessory is compatible with Agilent Instrumentation
£35.75 - £66.50
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
13406956
View Documents
Thermo Scientific™
60180-872
S/SL Injector Adaptor N/A N/A N/A
13496956
View Documents
Thermo Scientific™
60180-878
TCD Detector Adaptor N/A N/A N/A
13446956
View Documents
Thermo Scientific™
60180-873
Tight Nut N/A N/A N/A
Description
Description
QCC (Quick Column Change)-Leak Free Column Installation
  • Tool-free installation of analytical GC column
  • Graphite ferrules
Complete QCC Kits Includes:
  • Injector Adaptor
  • Detector Adaptor
  • Tight Nut
  • Ferrules (1 each 0.25, 0.32,0.53)
  • Ceramic Column Cutter
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ QCC (Quick Column Change) for Agilent Instruments