Learn More
Epredia™ Shandon™ Plastic Coverplates
For use on Shandon Sequenza™ and Cadenza™ Immunostainers
Brand:  Epredia™ 72110017
140.63 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
 View more versions of this product
Product Code. 12766807
  • £205.00 / Pack of 25
Explore more special offers
Description
Description
Saves on the use of costly reagents.
  • Device that fits over standard microscope slide allowing specimens to be immunostained with a minimum amount of reagent
  • Frees considerable technician time
  • Eliminates handling of slides between steps
  • Allows sections to be placed virtually anywhere on slide
  • Protects specimen from drying during long incubations
  • Ensures consistency in staining quality
Specifications
Specifications
Coverplate
25/Pk.