missing translation for 'onlineSavingsMsg'
Learn More
Learn More
This item is not returnable.
View return policy
Description
- Ready-to-use mucoliquefying preservative
- Liquefies (with agitation) mucoid cytology specimens
- Maintains cellular detail
- Short-term preservation of specimens
- Histology
- Cytology

Specifications
Light Red | |
4 x 3.8L (1 gal.) | |
Liquid |
Tissue Preservative | |
Provides short term preservation of specimens |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- 3.79 LT FIXATIVE, MUCOLEXX .
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 1B
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H350-May cause cancer.
- H411-Toxic to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
- EUH210-Safety data sheet available on request.
Precautionary Statement
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P321-Specific treatment (see on this label).
- P391-Collect spillage.
Supplemental information
- MIXTURE LIST-Contains : Ethanol, Lithium chloride, Formaldehyde,methanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Epredia™ Shandon™ Mucolexx™ > 3.8L x 1
Spot an opportunity for improvement?Share a Content Correction