missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SL1 Plus Centrifuge Series

Ergonomic design, standard high contrast user interface for up to 6 saved programs with simple push-button operation for routine applications.

Brand:  Thermo Scientific™ 75009000


3356.22 GBP valid until 2024-07-30
Use promo code "21657" to get your promotional price.



Additional Details : Weight : 44.00000kg

 View more versions of this product

Product Code. 17140579

  • £4580.00 / Each
Estimated Shipment: 25-09-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The SL1/SL1R Plus series centrifuges come standard with an easy-to-use keypad and high contrast LCD display.  Quickly and securely swap between 14 unique rotor options with the tool-less Auto-Lock rotor exchange. This compact centrifuge has a capacity up to 1.6L and can spin up to 76 x 5/7 mL blood tubes and 16 x 50mL conical tubes with biocontainment and is ideal for routine applications.

  • Easy to Use:

    • Intuitive controls simplify operation and provide detailed information on a wide range of operating parameters and processing status for optimal performance.
    • Modern industrial design supports continuous improvement in rotor loading ergonomics, user safety and current needed certifications
    • Motorized lid latch enables one finger downward motion for easiest closing and locking of centrifuge lid, even on tall benches.

    Thermo Scientific™ Auto-Lock™ III Rotor System:

    • Secure locking system allows easy push-button installation and exchange of rotors.
    • System saves time when switching between applications.
    • Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life.

    Thermo Scientific™ ClickSeal™ Bucket Sealing System:

    • Biocontainment solution seals with a snap; eliminates screw caps and complicated clips.
    • System has glove-friendly, one-handed open/close capability.
    • System is certified by CAMR™ in Porton Down, UK.

    Thermo Scientific™ SMARTSpin™ Technology:

    • Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option:
    • Technology optimizes safety and improves separations.

    Space-Saving Design:

    • Compact dimensions optimize work area and ergonomic height and curved design ensure easy sample loading and unloading and easy cleaning.

    Energy Efficient:

    • System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Specifications

Specifications

EN/UL/IEC 61010-1, EN/UL/IEC 61010-2-020 and 61010-2-101, EN/UL/IEC 61326-1, EN/UL/IEC 61326-2-6, FCC part 15,ISO 14971, ISO 13485, ISO 9001
4 x 400 mL with TX-400 Rotor
15,200 rpm (Microliter 30 x 2 rotor)
LED
Energy efficient, less waste
stores up to 6 programs
9 Accel/10 Braking
Each
SMARTSpin Imbalance detection, finger-pinch prevention, crash-proof construction
Yes
100/115/220/230 V
Microprocessor
25,830 x g (Microliter 30 x 2 Rotor)
No
Direct, Brushless Induction Low Profile
rotor needs to be purchased separately
IVD certified
Up to 6 programs via push button and high contrast LCD interface
No
General Purpose Centrifuge
14.2 x 17.5 x 26 in. (36.2 x 56.6 x 66 cm)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Services