Learn More
Thermo Scientific™ Sorvall™ Adapters for Stratos™ Four-Place Swinging Bucket Rotor
For use with Thermo Scientific Sorvall Stratos Centrifuges in Four-Place Swinging Bucket Rotor 75-003-047 with Round Bucket 75-008-172
Marca: Thermo Scientific™ 75008187
Code : 51
Dettagli aggiuntivi : Weight : 0.50000kg
Descrizione
- Accommodate tube sizes from 1.5 to 100mL, including urine tubes
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
Specifica
4 x 180 Swing bucket rotor | |
Brown |