Learn More
Thermo Scientific™ Tips for Tecan™ and Axon™ Instruments PatchXpress Workstations
GREENER_CHOICE
For use with Tecan™ Genesis™, Freedom Evo™, Cavro™, PatchXpress Workstations
Brand:  Thermo Scientific™ 902-262XP
91.19 GBP valid until 2025-12-31
Use promo code "24289" to get your promotional price.
 View more versions of this product
Product Code. 11510205
  • £116.00 / Pack of 960
Estimated Shipment: 11-08-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
For use with Tecan™ Genesis™, Freedom Evo™, Cavro™, PatchXpress Workstations
Clear pipet tips, Tecan style.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
50 μL
Non-sterile
Autoclavable
Rack
Tecan Genesis, Tecan Freedom EVO, Cavro, patchXpress
Filtered
White
DNase, RNase, and Pyrogen free
Automation
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Tips for Tecan™ and Axon™ Instruments PatchXpress Workstations > 50μL; Non sterile; Fits Tecan Genesis, Freedom Evo, Cavro