missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSX Series High-Performance Lab Refrigerators
BIOPHARMA

Product Code. 16337181 Shop All Thermo Scientific Products
Click to view available options
Capacity (English):
11.5 cu. ft.
23 cu. ft.
29.2 cu. ft.
45.8 cu. ft.
51.1 cu. ft.
Door Style:
Double Glass
Double Solid
Glass
Sliding Glass
Solid
Plug Type:
CEE 7/7
NEMA 6-15
Electrical Requirements:
208-230V, 50/60Hz
208-230V, 50Hz
208-230V, 60Hz
Unit Size:
Each
This item is not returnable. View return policy
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.

Product Code. 16337181

Brand: Thermo Scientific™ TSX1205SV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. Se returpolicy
Tillgänglighet och beräknat leveransdatum gäller standardutrustning. Om fabriksmonterade tillval, anpassningar eller tjänster krävs, vänligen begär en offert eller kontakta din kundansvarige, eftersom leveranstiderna kan variera.

Innovative refrigeration solutions for healthcare and research needs.

Thermo Scientific™ TSX Series high-performance lab refrigerators are designed with features that support sample protection and sustainability objectives for the storage of media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade storage requiring 3° to 7°C. The Series' V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

Featuring over 450W of reserve refrigeration capacity, the TSX series has the power to protect your product from temperature swings as a result of frequent and prolonged door access, large product loads, and other activities that introduce hear to the cabinet.

The TSX Series delivers the performance and quality you need, in addition to the low sound, low energy, and exceptional user experience you deserve.

The drive for sample integrity and security

  • Designed to safely store media, reagents, chemotherapy, pharmaceutical and other medical and laboratory-grade products requiring a temperature range of 3°C to 7°C
  • Unique V-drive technology is designed to detect usage patterns such as door openings when a higher compressor speed is needed and periods of stability where the compressor runs at a lower speed, saving energy without compromising protection
  • Easy to use microprocessor control with interior light, setpoint security and alarms
  • Standard door locks provide comfort and assurance

Enabling compliance

  • Tested to GMP Clean Room Class A / ISO 5 & 6 (ISO EN 14644-1) with easy to clean surfaces support USP <797> and <800> hazardous drug compounding and other clean room environments
  • Tested and certified to conform with the design and safety standards of UL 471, 61010, and CE Directive 2011/65/EU and 2015/863

Designed for an ergonomic and comfortable experience

  • Ergonomically designed door handle with a soft-touch grip
  • The controller is angled to help prevent neck strain
  • Convenient, easy to clean and adjustable epoxy coated full and half shelves
  • Whisper quiet with a sound level of 52 dBA, enabling end users to work alongside the refrigerator without distraction
  • Bright, LED interior lighting
  • Self-closing multipaned glass or solid doors with 90° stop to assist with inventory loads

Verified sustainability

  • ACT Environmental Impact Factor Label Certified, providing third-party verification of the sustainable impacts of the product, its operations, and its end of life. **
  • Natural, R290 refrigerants and water-blown foam make the TSX Series compliant with EU F-GAS regulations, U.S. Environmental Protection Agency's Significant New Alternatives Policy (SNAP), and other Global Environmental Regulations.
  • Manufactured in an award-winning, zero-waste to landfill facility

Ready for connectivity and monitoring

  • Thermo Scientific™ DeviceLink™ Connect ready, enabling you to have a connected experience with your unit so you always—and anywhere— know the safety of your samples and the performance of your refrigerator or freezer.
  • Compatible with wireless monitoring systems and remote alarm contacts including our Thermo Scientific™ Smart-Vue™ Pro wireless monitoring systems

Make it your own

  • Alternate Door swing configuration and door types
  • Extra and Alternate wire baskets, shelves, and drawers
  • Free-standing or factory-installed chart recorders
  • Stainless Steel interior finish
  • E-Lock Adapter kits
TRUSTED_SUSTAINABILITY

Specifikationer

Type High-Performance Refrigerator
Capacity (English) 11.5 cu. ft.
Capacity (Metric) 326 L
Temperature (Setpoint) 3°-7°C (5°C)
Refrigerant R290
Door Style Solid
No. of Doors 1
Shelves 4
Certifications/Compliance UL, cUL, CE
Display Microprocessor
Product Line TSX Series
Defrost Automatic
Monitoring Options Alarm, Recorder
Chart Recorder Optional
Smart View Compatibility Smart-Vue Compatible
Dimensions (DxWxH) Exterior 31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Dimensions (D x W x H) Interior 21.8 x 20.0 x 52.4 in. (55.4 x 50.8 x 133.1 cm)
Dimensions (L x W x H) Exterior 31.1 x 24.0 x 73.0 in. (79.0 x 61.9 x 185.4 cm)
Depth (English) Exterior 31.1 in.
Depth (English) Interior 21.8 in.
Depth (Metric) Exterior 79.0 cm
Depth (Metric) Interior 55.4 cm
Height (English) Exterior 73.0 in.
Height (English) Interior 52.4 in.
Height (Metric) Exterior 185.4 cm
Height (Metric) Interior 133.1 cm
Width (English) Exterior 24.0 in.
Width (English) Interior 20 in.
Width (Metric) Exterior 61.9 cm
Width (Metric) Interior 50.8 cm
Amperage 15 A
Plug Type CEE 7/7
Voltage 230 V
Electrical Requirements 208-230V, 50Hz
Frequency 50/60 Hz
Shipping Weight (English) 340 lb.
Shipping Weight (Metric) 154 kg
Weight (English) 286 lb.
Weight (Metric) 130 kg
Visa mer Visa mindre

*Data on file. Based on comparisons between TSX Series and previous generation Thermo Scientific models.

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.