Learn More
Thermo Scientific™ Tuf-Bond™ PTFE/Silicone Discs
Discs that combine the inertness of PTFE with the resealability of silicone
Brand:  Thermo Scientific™ TS-12720
Product Code. 11787047
  • £162.00 / Pack of 72
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Structurally bonded PTFE to silicone; no cement to leak out of your sample after needle penetration
  • Reseals instantly, puncture after puncture
  • Autoclavable
  • Compresses to maintain a tight seal forcing the PTFE to conform to the sealing surface
  • Standard syringe and GC needles penetrate the entire disc with ease
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE, Silicone