missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Ultra Disposable Microtome Blades

Epredia™ ultra microtome blades offer high consistency, impressive durability, dependability, and precise performance.

Brand:  Epredia™ 3053835

98.40 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.


 View more versions of this product

Product Code. 12191830

  • £181.00 / Pack of 50

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High- and low-profile universal blades can be used with any microtome or cryostat on the market
  • Unique angle profile that facilitates faster ribboning
  • Increased Longevity: Produces an edge that is twice as strong as traditional blades
  • Reliable Consistency: New proprietary process ensures a durable and consistent coating

Specifications

Specifications

Low
Microtomes, Cryostats
Single-use
34°
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.