Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation Documention
Product Code. 16275342 Shop All Thermo Scientific Products
Product Code. 16275342
missing translation for 'mfr': Unity Lab Services™ IOQDOCE89003810
OEM service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific Ultra-Low Temperature Freezers Models: Thermo Scientific TSX-series (post 2018), Forma 89000-series, HeraFreeze HLE, Revco RLE, Thermo Scientific TLE
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Thermo Scientific Ultra-Low Temperature Freezers including all Thermo Scientific Ultra-Low Temperature Freezers Models: Thermo Scientific TSX-series (post 2018), Forma 89000-series, HeraFreeze HLE, Revco RLE, Thermo Scientific TLE
Description Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing.
Location Contact your local sales representative for a list of standard and extended coverage areas.
Includes IQ/OQ Documentation