Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation Service
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Ultra-Low Temprature Freezers Models: Thermo Scientific TSX-series (post 2018), Forma 89000-series, HeraFreeze HLE, Revco RLE, Thermo Scientific TLE
Brand: Unity Lab Services™ IOQPCKE89003810
Additional Details : Weight : 0.00010kg
Description
IQ
- Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
- Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ System Component Information Detailed configuration information for each system component have been recorded.
- OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
- Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment.
- Key Tests: Door Ajar, Power Failure, Temperature Alerts
Specifications
Thermo Scientific Ultra-Low Temperature Freezers including Ultra-Low Temprature Freezers Models: Thermo Scientific TSX-series (post 2018), Forma 89000-series, HeraFreeze HLE, Revco RLE, Thermo Scientific TLE | |
Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing. | |
Contact your local sales representative for a list of standard and extended coverage areas. | |
Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service |