Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation Service with Temperature Map
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Ultra-Low Temprature Freezers Models: Thermo Scientific TSX-series (post 2018), Forma 89000-series, HeraFreeze HLE, Revco RLE, Thermo Scientific TLE
Brand: Unity Lab Services™ IOPQPCKE89003810
2894.25 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
This item is not returnable.
View return policy
Description
IQ
- Unpacking, Assembly and Installation Verifies that the equipment, manuals, supplies and any other accessories arrived undamaged as specified in the sales order. Verifies equipment is assembled and installed and any initial diagnostics tests have been performed. Any abnormal event(s) observed during assembly and installation have been documented.
- Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
OQ
- System Component Information Detailed configuration information for each system component have been recorded.
- OQ Limits: Record a list of manufacturer recommended limits for ensuring that the system is operating as expected.
- Equipment Operational Tests and Results: Test important equipment functions to verify that the equipment operates as intended by the manufacturer and required by the user. This is a group of selected important equipment parameters for testing according to the intended use of the equipment. - Key Tests: Door Ajar, Power Failure, Temperature Alerts
Temp Map
- Temperature Mapping Tests and Results: Test important equipment capabilities for ensuring that the equipment is performing in accordance with the intended use of the system.
Multi-Point Temperature Mapping for a 24 hour period with 1 minute data intervals.
Specifications
Thermo Scientific Ultra-Low Temperature Freezers including Ultra-Low Temperature Freezers Models: Thermo Scientific TDE, Forma FDE, Revco RDE, Isotemp IUE | |
Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing. Temperature Mapping: 24 hours of multi-point mapping. | |
Contact your local sales representative for a list of standard and extended coverage areas. | |
Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service |