Learn More
Unity Lab Services™ Thermo Scientific™ Ultra-Low Temperature Freezers Validation and Temperature Map Documention
Original equipment manufacturer (OEM) service provider for Thermo Fisher Scientific. All protocols designed to optimize efficiency are included with each Ultra-Low Temperature Freezers Models: Thermo Scientific TSX-series, Forma 89000-series, HeraFreeze HLE, Revco RLE, Thermo Scientific TLE
Brand: Unity Lab Services™ IOPQDOCE89003810
Additional Details : Weight : 0.45000kg
Description
IQ/OQ/Temperature Mapping documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept upto date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.Specifications
Thermo Scientific Ultra-Low Temperature Freezers including Ultra-Low Temperature Freezers Models: Thermo Scientific TSX-series (post 2018), Forma 89000-series, HeraFreeze HLE, Revco RLE, Thermo Scientific TLE | |
Documentation Only for the following, Services Not included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform empty chamber temperature mapping, open door and power loss testing. Temperature Mapping: 24 hours of multi-point mapping. | |
Contact your local sales representative for a list of standard and extended coverage areas. | |
IQ/OQ/Temp Map Documentation |