Learn More
Unity Lab Services™ Thermo Scientific™ Water Purification Validation Documention for Thermo Scientific™ Barnstead LabTower II
As an original equipment manufacturer (OEM) service provider for Thermo Fisher Scientific, all protocols designed to optimize efficiency are included with each Thermo Scientific Water Purification Validation Documention for RThermo Scientific Barnstead LabTower II
Brand: Unity Lab Services™ IOQDOCE50134156
Additional Details : Weight : 50.00000kg
Description
IQ/OQ documentation that supports regulatory requirements and industry standards for instrument compliance. Our documents are kept up to date by conducting annual reviews. We work directly with our product experts to have the most updated documentation available.
Specifications
Thermo Scientific Water Purification including Thermo Scientific Barnstead LabTower II | |
Documentation Only for the following, Services Not Included: Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, Flow Adjustment, Water Pressure, Resistivity, Power Failure testing. | |
Contact your local sales representative for a list of standard and extended coverage areas. | |
IQ/OQ Documentation |