Learn More
Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels
Store and dispense small amounts of liquid nitrogen with our economical and durable liquid nitrogen transfer vessels.
Brand: Thermo Scientific™ TY509X3
1235.83 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
Store and dispense small amounts of liquid nitrogen with our economical and durable Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels. The Thermo Series includes six models with capacities from 5 to 32 L. A full range of handling and dispensing accessories as well as transport carts are also available.
- Designed to bring liquid nitrogen to other cryovessels
- Lightweight aluminum dewars assure low-static evaporation rates
- Optional withdrawal device, 12mL dipper and wheeled accessory cart
- Compact Thermo 5 and 10 dewars feature a convenient pail-style handle for pouring and for use in applications where only small quantities of liquid nitrogen are needed
- Thermo 10, 20, and 30 can be fitted with self-pressurized withdrawal accessory devices to easily dispense LN2 without pouring
Note:
Stated static evaporation rates are based on new container performance with no lid openings and no product loads. Actual working performance may vary with individual applications.
Specifications
LN2 Transfer Vessel | |
24.7 in. | |
14.5 in. | |
36.8 cm | |
24 lb. | |
8.6 kg |
20 L | |
62.8 cm | |
2 in. | |
5 cm | |
19 lb. |