Learn More
Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels
Store and dispense small amounts of liquid nitrogen with our economical and durable liquid nitrogen transfer vessels.
£1279.87 - £1720.61
Specifications
Type LN2 Transfer Vessel
View More Specs
Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10480394
View accessory products View Documents Promotion Details
Thermo Scientific™
TY509X1
5 L
£1279.87
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
10318543
View accessory products View Documents Promotion Details
Thermo Scientific™
TY509X2
10 L
£1353.18
Each
Estimated Shipment: 29-04-2025
to see stock.
Add to basket
10398293
View accessory products View Documents Promotion Details
Thermo Scientific™
TY509X3
20 L
£1500.11
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
11587304
View accessory products View Documents Promotion Details
Thermo Scientific™
TY509X4
32 L
£1720.61
Each
Estimated Shipment: 02-05-2025
to see stock.
Add to basket
Description
Description
Store and dispense small amounts of liquid nitrogen with our economical and durable Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels. The Thermo Series includes six models with capacities from 5 to 32 L. A full range of handling and dispensing accessories as well as transport carts are also available.
Features:
  • Designed to bring liquid nitrogen to other cryovessels
  • Lightweight aluminum dewars assure low-static evaporation rates
  • Optional withdrawal device, 12mL dipper and wheeled accessory cart
  • Compact Thermo 5 and 10 dewars feature a convenient pail-style handle for pouring and for use in applications where only small quantities of liquid nitrogen are needed
  • Thermo 10, 20, and 30 can be fitted with self-pressurized withdrawal accessory devices to easily dispense LN2 without pouring
Note:
Stated static evaporation rates are based on new container performance with no lid openings and no product loads. Actual working performance may vary with individual applications.
  • Warranty varies by country, please contact us for details
Specifications
Specifications
LN2 Transfer Vessel
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels