Learn More
Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple
Use these probes for a variety of high-temperature applications. Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple Probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
Brand:  Cole-Parmer™ 08505-85
 View more versions of this product
Product Code. 11706624
  • £121.00 / Pack of 6
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Flexible Kapton-insulated wire is ideal for checking food temperatures
  • Flexible fiberglass-Insulated wire used for high-temperature measurements
  • Custom designs or changes to this product are available upon request
  • Exposed-junction probes are not recommended for use with aqueous solutions
  • Do not subject type J exposed-junction probes to moisture as they will rust
Specifications
Specifications
Thermocouple Probe
Ideal for Checking Food Temperatures
Kapton
60 in. (probe)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple > Kapton-Insulated; Blue; Temperature Range: -150 to 260deg.C