Learn More
Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple
Use these probes for a variety of high-temperature applications. Cole-Parmer™ Digi-Sense™ Insulated-Wire Thermocouple Probes include straight insulated cable with no handle and feature ANSI color-coded mini connectors: type J - black, type K - yellow, and type T - blue.
Brand:  Cole-Parmer™ 08512-82
 View more versions of this product
Product Code. 11841280
  • £44.70 / Each
Estimated Shipment: 10-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Flexible Kapton-insulated wire is ideal for checking food temperatures
  • Flexible fiberglass-Insulated wire used for high-temperature measurements
  • Custom designs or changes to this product are available upon request
  • Exposed-junction probes are not recommended for use with aqueous solutions
  • Do not subject type J exposed-junction probes to moisture as they will rust
Specifications
Specifications
-150°C to +482°C
Thermocouple Probe
Used for High-Temperature Measurements
120 in. (probe)