Learn More
HUBER Thermofluid Ethylenglykol
For use with chillers
Brand: HUBER 6170
Code : 04
Additional Details : Weight : 12.20000kg
Description
Thermofluid - Ethylenglykol, for ChillersSpecifications
10 L | |
Chillers | |
Thermofluid Ethylenglykol | |
10 L |
Safety and Handling
- Monoethylenglykol
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P314-Get medical advice/attention if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501-Dispose of contents/container to .