missing translation for 'onlineSavingsMsg'
Learn More

HUBER Thermofluid Ethylenglykol

For use with chillers

Brand:  HUBER 6171

Code : 04

Additional Details : Weight : 50.00000kg


 View more versions of this product

Product Code. 11597354

  • £1653.60 / 50 L
Estimated Shipment: 02-09-2024
to see stock.
Add to basket

Description

Description

Thermofluid - Ethylenglykol, 50L, for Chillers
Specifications

Specifications

50 L
Chillers
Thermofluid Ethylenglykol
50 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • MEG, Monoethylenglykol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H302-Harmful if swallowed.
  • H373-May cause damage to organs.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ethanediol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers