Learn More
Thermo Scientific™ Thermolyne™ Benchtop Muffle Furnaces
Ideal for general laboratory use, including gravimetric analysis, sintering, quantitative analysis and heat treating.
£3028.92
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10509722
|
Thermo Scientific™
F48010-33 |
5 L |
£3028.92
Each |
Estimated Shipment: 16-08-2024
Log in to see stock. |
|||||
Description
Ideal for general laboratory use, including gravimetric analysis, sintering, quantitative analysis and heat treating.
- For added protection, the door safety switch stops power to heating elements when door opens
- Thermocouple break protection cuts power to heating elements, preventing a thermocouple failure runaway condition
- Two open coil heating elements on chamber sides assure fast heat-up with minimum temperature gradient
- Thermal-efficient ceramic insulation surrounds chamber for maximum energy efficiency
- 0.95cm (0.38 in.) dia. port for monitoring chamber temperatures with independent measuring device at rear of chamber
F47900 Models
- 2L (0.07 cu. ft.) of heating area
F48000 Models
- 5.9L (0.20 cu. ft.) of heating area
- Includes ceramic shelf (SH480X1) that doubles furnace load capacity
A1 Controller
- Digital single setpoint control
- Single display shows actual temperature or setpoint
B1 Controller
- Digital single setpoint control with a single ramp to setpoint and a dwell
- Single display shows actual temperature or setpoint
- Furnaces with this control also use a mechanical Overtemperature Protection relay
C1 Controller
- Digital programmable control with one stored program of eight segments
- Furnaces with this control also use a mechanical Overtemperature Protection relay
D1 Controller
- Digital programmable control with four stored programs, 16 segments per program
- With RS-232 communications interface
- Furnaces with this control also use a mechanical Overtemperature Protection relay
Includes:
Power cord (-60 models include 20-amp cord; -33 units available with plug for China)
Warranty:
1 year (parts and labor)
Specifications
100°C to 1200°C |