missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ F1.5 ThermoMixer

The perfect choice for mixing and heating established routine applications, with a fixed block for 1.5mL tubes and plates.

Brand:  Eppendorf™ 5384000039

1994.43 GBP valid until 2024-12-06
Use promo code "21633" to get your promotional price.


Product Code. 15189752

  • £2811.20 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Reliable prevention of condensate formation in small vessels can be achieved with the heated lid
  • Up to 1,500 rpm for efficient sample mixing
  • Outstanding mix control technology results in excellent mixing performance
  • Anti-spill technology prevents lid wetting and cross-contamination
  • Predefined temperature keys (37°, 42°, 56°, and 95°C) for simple and intuitive operation
  • Very fast heat up speed of up to 18°C/min, reducing your waiting time
  • Ergonomic design and operation
  • Applications include growth of baceria and yeasts, enzymatic reactions, transformation of bacteria strains and plasmids, denaturation of DNA, RNA and proteins, lysis reactions at 100°C, labeling of nucleic acids and proteins, and more
TRUSTED_SUSTAINABILITY
Specifications

Specifications

±0.5°C at 20° to 45°C
Thermoblock For 24 Reaction Vessels 1.5 Ml
220 to 240 V, 50 to 60 Hz, 200 W
170 mm
304 mm
50 to 60 Hz
200 W
1500 rpm
4 to 100°C
220 to 240 V
ThermoMixer
206 mm
Transformation of Bacteria Strains and Plasmids
4°C to 100°C
1.5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers