missing translation for 'onlineSavingsMsg'
Learn More

Watson-Marlow Thermoplastic Marprene Tubing

Use this tubing for a wide variety of applications. Watson Marlow™ Thermoplastic Marprene Tubing is resistant to a wide range of chemicals, offers long tubing life and is highly resistant to oxidizing agents such as ozone, peroxides and sodium hypochlorite.

Brand:  Watson-Marlow 902.0032.016

Code : 50

Additional Details : Weight : 0.46080kg

Product Code. 10488501

  • £524.00 / 15m
In Stock
to see stock.

Description

Description

  • Opaque beige tubing
  • Fair spallation
  • Certification USP Class VI, FDA regulations 21 CFR177.2600 for contact with food
  • Operating temperature -5 to +80°C
  • Available in thirty different bore sizes, five wall thicknesses and three coil lengths including bulk reels
  • Marprene may be steam sterilized at 135°C or cold sterilized by ethylene oxide.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers