missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Thiamine hydrochloride, 98.5-101.5% Product Code.: 10477540

Thermo Scientific Chemicals Thiamine hydrochloride, 98.5-101.5%

Product Code. 10477540
10 g, Glass bottle
Click to view available options
Quantity:
10 g
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10477540

Brand: Thermo Scientific Chemicals 148990100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Thiamine hydrochloride, 98.5-101.5%, C12H17ClN4OS.HCl, CAS Number-67-03-8, aneurine hydrochloride, berin, thiamin chloride, thiamine chloride hydrochloride, thiamine dichloride, thiamine hcl, thiamine hydrochloride, thiaminium chloride, trophite, vitamin b1 hydrochloride, 1kg, 98.5 to 101.5%

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H17ClN4OS·HCl
Molecular Weight (g/mol) 337.26
MDL Number MFCD00012780
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
Synonym thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chlorideShow MoreShow Less
PubChem CID 6202
ChEBI CHEBI:49105
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
SMILES CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]

Specifications

Melting Point 250.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5%
Packaging Glass bottle
Quantity 10 g
Beilstein 27, IV, 1766
Merck Index 15, 9447
Solubility Information Solubility in water: 1 g/mL. Other solubilities: 1 g/18 mL glycerol - 1 g/100 mL 95% alcohol, 1 g/315 mL abs. alcohol, soluble in propylene glycol, practically insoluble in hexane, chloroform, insoluble in benzene, diethyl ether
Formula Weight 337.26
Percent Purity 98.5-101.5%
Physical Form Crystalline Powder
Chemical Name or Material Thiamine hydrochloride, 98.5-101.5%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453529 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number148990010,148991000,148990100
Lot NumberA0429851 Certificate TypeCertificate of Analysis Date06/05/2024 Catalog Number148990010,148991000,148990100
Lot NumberA0428392 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number148990010,148991000,148990100
Lot NumberA0443248 Certificate TypeCertificate of Analysis Date18/12/2023 Catalog Number148990010,148991000,148990100
Lot NumberA0453529 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number148990010,148991000,148990100
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Thiamine hydrochloride, 98.5-101.5% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.