missing translation for 'onlineSavingsMsg'
Learn More
Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals Product Code.: 11483930

Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals

Product Code. 11483930
100 g
Click to view available options
Quantity:
100 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11483930

Brand: Thermo Scientific Alfa Aesar A19560.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thiamine hydrochloride plays an important role as a nutrient for carbohydrate metabolism and in nerve function. It finds application as a food-additive to add a brothy/meaty flavor to gravies or soups. It is involved in the preparation of 3-(4-amino-2-methyl-pyrimidin-5-ylmethyl)-5-(2-hydroxy-ethyl)-4-methyl-thiazolium chloride. In addition, it finds use in amino acid synthesis in cultured cells, which is present in many classical and serum-free formulations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiamine hydrochloride plays an important role as a nutrient for carbohydrate metabolism and in nerve function. It finds application as a food-additive to add a brothy/meaty flavor to gravies or soups. It is involved in the preparation of 3-(4-amino-2-methyl-pyrimidin-5-ylmethyl)-5-(2-hydroxy-ethyl)-4-methyl-thiazolium chloride. In addition, it finds use in amino acid synthesis in cultured cells, which is present in many classical and serum-free formulations.

Solubility
Soluble in water, glycerol, ethanol, dimethyl sulfoxide and methanol. Insoluble in benzene, hexane, chloroform and ether.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-03-8
Molecular Formula C12H18Cl2N4OS
Molecular Weight (g/mol) 337.263
MDL Number MFCD00012780
InChI Key DPJRMOMPQZCRJU-UHFFFAOYSA-M
Synonym thiamine hydrochloride, vitamin b1 hydrochloride, thiamine hcl, aneurine hydrochloride, berin, thiamine chloride hydrochloride, trophite, thiamin chloride, thiamine dichloride, thiaminium chlorideShow MoreShow Less
PubChem CID 6202
ChEBI CHEBI:49105
IUPAC Name 2-[3-[(4-amino-2-methylpyrimidin-5-yl)methyl]-4-methyl-1,3-thiazol-3-ium-5-yl]ethanol;chloride;hydrochloride
SMILES CC1=C(SC=[N+]1CC2=CN=C(N=C2N)C)CCO.Cl.[Cl-]

Specifications

Melting Point ∼250°C (decomposition)
Assay Percent Range 0.99
Quantity 100 g
Beilstein 3851771
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,9295
Solubility Information Soluble in water,glycerol,ethanol,dimethyl sulfoxide and methanol. Insoluble in benzene,hexane,chloroform and ether.
Formula Weight 337.28
Percent Purity 99%
Assay (dry wt.), may cont. up to 5% water
Chemical Name or Material Thiamine hydrochloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberW26F007 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog NumberA19560.22,A19560.0B,A19560.30
Lot NumberU14F057 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog NumberA19560.22,A19560.0B,A19560.30
Lot NumberM21H022 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog NumberA19560.22,A19560.0B,A19560.30
Lot Number10237472 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog NumberA19560.22,A19560.0B,A19560.30
Lot NumberN08K004 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog NumberA19560.22,A19560.0B,A19560.30
compliance-icons
Product Identifier
  • Thiamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Thiamine hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thiamine hydrochloride, 99% (dry wt.), may cont. up to 5% water, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.