missing translation for 'onlineSavingsMsg'
Learn More

Thiazole-5-methanol, 95%, Thermo Scientific™

£29.50 - £90.99

Chemical Identifiers

CAS 38585-74-9
Molecular Formula C4H5NOS
Molecular Weight (g/mol) 115.15
InChI Key WKBQQWDVVHGWDB-UHFFFAOYSA-N
Synonym 5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl-1,3-thiazole
PubChem CID 2763216
IUPAC Name 1,3-thiazol-5-ylmethanol
SMILES C1=C(SC=N1)CO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15335018
View Documents Promotion Details
Thermo Scientific Acros
455550050
5 g
£29.50
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15345018
View Documents Promotion Details
Thermo Scientific Acros
455550250
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Chemical Identifiers

38585-74-9
115.15
5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl-1,3-thiazole
1,3-thiazol-5-ylmethanol
C4H5NOS
WKBQQWDVVHGWDB-UHFFFAOYSA-N
2763216
C1=C(SC=N1)CO

Specifications

38585-74-9
94% min. (GC)
1.32
WKBQQWDVVHGWDB-UHFFFAOYSA-N
1,3-thiazol-5-ylmethanol
2763216
95%
1.3200g/mL
C4H5NOS
5-hydroxymethylthiazole, 5-hydroxymethyl thiazole, thiazol-5-ylmethanol, 5-thiazolemethanol, thiazole-5-methanol, thiazol-5-yl-methanol, thiazol-5-yl methanol, 5-thiazolylmethanol, 5-hydroxymethyl-1,3-thiazole
C1=C(SC=N1)CO
115.15
115.15
Thiazole-5-methanol
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes serious eye damage. Harmful to aquatic life with long lasting effects.

GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

Product Identifier
  • Thiazole-5-methanol

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only