missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thieno[2,3-b]pyrazine-6-carboxylic acid, 90%, Thermo Scientific™
£72.55 - £181.00
Chemical Identifiers
CAS | 59944-79-5 |
---|---|
Molecular Formula | C7H4N2O2S |
Molecular Weight (g/mol) | 180.181 |
InChI Key | MEHCDDSVVYRWJT-UHFFFAOYSA-N |
Synonym | thieno 2,3-b pyrazine-6-carboxylic acid, pubchem9882, thieno 2,3-b pyrazine-6-carboxylicacid, 6-thieno 2,3-b pyrazinecarboxylic acid, thieno 3,2-b pyrazine-6-carboxylic acid, thiopheno 2,3-b pyrazine-6-carboxylic acid |
PubChem CID | 7164578 |
IUPAC Name | thieno[2,3-b]pyrazine-6-carboxylic acid |
SMILES | C1=CN=C2C(=N1)C=C(S2)C(=O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10167992
|
Thermo Scientific Maybridge
CC31501CB |
250 mg | Amber Glass Bottle |
£72.55
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10177992
|
Thermo Scientific Maybridge
CC31501DA |
1 g | Amber Glass Bottle |
£181.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
59944-79-5 | |
180.181 | |
thieno 2,3-b pyrazine-6-carboxylic acid, pubchem9882, thieno 2,3-b pyrazine-6-carboxylicacid, 6-thieno 2,3-b pyrazinecarboxylic acid, thieno 3,2-b pyrazine-6-carboxylic acid, thiopheno 2,3-b pyrazine-6-carboxylic acid | |
thieno[2,3-b]pyrazine-6-carboxylic acid |
C7H4N2O2S | |
MEHCDDSVVYRWJT-UHFFFAOYSA-N | |
7164578 | |
C1=CN=C2C(=N1)C=C(S2)C(=O)O |
Specifications
59944-79-5 | |
C7H4N2O2S | |
MEHCDDSVVYRWJT-UHFFFAOYSA-N | |
thieno[2,3-b]pyrazine-6-carboxylic acid | |
7164578 | |
90% |
90% | |
thieno 2,3-b pyrazine-6-carboxylic acid, pubchem9882, thieno 2,3-b pyrazine-6-carboxylicacid, 6-thieno 2,3-b pyrazinecarboxylic acid, thieno 3,2-b pyrazine-6-carboxylic acid, thiopheno 2,3-b pyrazine-6-carboxylic acid | |
C1=CN=C2C(=N1)C=C(S2)C(=O)O | |
180.181 | |
180.19 | |
Thieno[2, 3-b]pyrazine-6-carboxylic acid |