missing translation for 'onlineSavingsMsg'
Learn More

Thieno[2,3-b]pyridine-2-carbaldehyde, ≥97%, Thermo Scientific™

Catalog Number 10371043
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
53174-98-4
C8H5NOS
163.194
MFCD07772791
HGZQKYOLRLVMHZ-UHFFFAOYSA-N
thieno 2,3-b pyridine-2-carbaldehyde, thieno 2,3-b pyridine-2-carboxaldehyde, 2-formylthieno 2,3-b pyridine, thiopheno 2,3-b pyridine-2-carbaldehyde
289925
thieno[2,3-b]pyridine-2-carbaldehyde
C1=CC2=C(N=C1)SC(=C2)C=O
This item is not returnable. View return policy
53174-98-4
C8H5NOS
163.194
MFCD07772791
HGZQKYOLRLVMHZ-UHFFFAOYSA-N
thieno 2,3-b pyridine-2-carbaldehyde, thieno 2,3-b pyridine-2-carboxaldehyde, 2-formylthieno 2,3-b pyridine, thiopheno 2,3-b pyridine-2-carbaldehyde
289925
thieno[2,3-b]pyridine-2-carbaldehyde
C1=CC2=C(N=C1)SC(=C2)C=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 163.2
Percent Purity 97%
Chemical Name or Material Thieno[2, 3-b]pyridine-2-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'