Learn More
Faster™ Thimble Exhaust Connection for SafeFAST and CytoFAST Cabinets
Connection to allow cabinet to be connected to thimble ductwork.
£359.00 - £550.00
Specifications
For Use With (Application) Connecting Cabinet to Ductwork
View More Specs
Products 4
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
16278590
View Documents
Faster™
FX0002154070
SafeFAST Classic, Elite, Premium, 109, 215, 315
£499.00
Each
15523366
View Documents
Faster™
FX0002094070
SafeFAST Classic, Elite, Premium, CytoFAST Elite 209
£359.00
Each
15563366
View Documents
Faster™
FX0002124070
SafeFAST Classic, Elite, Premium, CytoFAST Elite 212
£477.00
Each
15583376
View Documents
Faster™
FX0002184070
SafeFAST Classic, Elite, Premium, CytoFAST Elite 112, 115, 218, 318
£550.00
Each
Description
Description
  • Specifically for SafeFAST Classic, Elite, Premium, CytoFAST Elite cabinets
Specifications
Specifications
Connecting Cabinet to Ductwork