Learn More
Thermo Scientific Chemicals Thin-Bore Sample Tube, 2mm ID
For use with MKII Magnetic Susceptibility Balances
£73.47 - £298.49
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11490400
|
Thermo Scientific Chemicals
014350.KG |
1 Each |
£73.47
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15441067
|
Thermo Scientific Chemicals
014350.KJ |
5 Pack |
£298.49
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- I.D.: 2mm
Specifications
Thin-bore Sample Tube |
Safety and Handling
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.