missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Thin Wall 0.5 mL PCR Tubes with Attached Caps

Manufactured without lubricants or chemical additives that can contaminate reagents and samples.Thin Wall 0.5mL PCR Tubes with Attached Caps are autoclavable and RNase-, DNase-, and DNA-free. Come non-sterile. Thin, uniform walls ensure consistent, precise thermal transfer. Attached flat caps form a tight seal to help prevent sample evaporation but open easily. Compatible with most thermal cyclers.

Brand:  BRAND™ 781310

40.20 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 10025720

  • £98.50 / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Manufactured from high-quality virgin plastics for excellent, reproducible results in most PCR applications, and are manufactured without lubricants or chemical additives that can contaminate reagents and samples.
  • Thin, uniform walls to ensure consistent, precise thermal transfer for excellent results.
  • Autoclavable (come non-sterile) and RNase-, DNase-, and DNA-free.
  • Compatible with most thermal cyclers.
  • Supplied with caps that form a tight seal and help prevent sample evaporation, yet are easy to open and close.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RNase, DNase and DNA free
0.5 mL
Thermal cyclers for excellent reproducible results in most PCR applications
Polypropylene
Autoclavable
Single-use
0.5 mL
Clear
1000 Pack
Non-sterile
Flat
Yes
Tube
Thin Wall PCR Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number24202 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog Number781310
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
BRAND™ Thin Wall 0.5 mL PCR Tubes with Attached Caps > 0.5mL; Clear

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.