missing translation for 'onlineSavingsMsg'
Learn More

Wilmad-LabGlass™ Thin Walled High Throughput NMR Tubes

Possess an average camber of 60 microns to guarantee the clarity of the spectrum for small molecule (MW<250) samples up to 600 MHz. Wilmad Thin Walled High Throughput NMR Tubes are designed for routine use in most low to mid field NMR spectrometers.

Brand:  Wilmad-LabGlass™ 66000-0061

 View more versions of this product

Product Code. 15398628

  • £245.00 / Pack of 100
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Camber and concentricity values listed are the Total Indicator Reading (TIR)
  • Designed for routine use in most low to mid field NMR spectrometers
  • One of the best OD tolerances in the industry
  • Guaranteed to fit tightly in spinner turbine
  • Made from ASTM E438 Type 1 Class B glass
  • 100% inspected for surface defects and physical dimension to ensure the success of your experiments
  • WG-1000-4-SJ and WG-1000-7-SJ are supplied with authentic Bruker SampleJet caps, all others come with a disposable NMR cap
Note: For NMR experiments that involve cooling, heating, biological sample, multi-dimension, multi-nuclei or DNP techniques, please refer to Wilmad Precision NMR Tubes.
Specifications

Specifications

60 μm
ASTM E438 Type 1 Class B Glass
MHz Rating: 600
5 mm
0.43 mm
WG-1000-4-SJ
103.5 mm
NMR Tube
Bruker SampleJet Cap
4 in.
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.