Learn More
Greiner Bio-One ThinCert™ Tissue Culture Inserts
Research tool for transport, co-culture, migration and invasion studies. Greiner Bio-One ThinCert™ Tissue Culture Inserts provides improved cell adhesion due to physical surface treatment.
Brand: Greiner Bio-One 657610
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Clear polystyrene housing with sealed PET capillary pore membrane
- Hanging geometry provides protection to lower compartment cell layer
- Simplified pipetting due to self-lift geometry
- Eccentric in-well positioning allows unequalled access to lower compartment
- Available in 6, 12 and 24 well format
Especificaciones
452.4 mm2 | |
2.0 to 4.15 mL | |
1.0 μm | |
6 |
Polystyrene | |
2 x 106 pores/cm2 | |
Sterile |