missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Thinline Slotted Cassettes in Tube Packs

Product Code. p-7193540 Shop All Epredia Products
Click to view available options
Color:
Beige
Blue
Gray
Green
Lavender
Light Green
Lilac
Orange
Pink
Tan
White
Yellow
Unit Size:
Pack of 750
This item is not returnable. View return policy

Product Code. 16384096

Brand: Epredia™ B851736BE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For routine tissue samples, cassettes are compatible with all Epredia PrintMate™ Cassette Printers and have a front lid hinge with color-matched, attached lids available in 12 different color varieties.

Printing

  • Surface finish and reinforced print faces are engineered to optimize print quality
  • Tube design is precisely engineered to reduce jamming and prevent downtime

Grossing, Processing and Embedding

  • Cassette design and flow characteristics tested to ensure efficient reagent exchange
  • Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples

Sectioning

  • Precision engineering for solid connection to microtomes

Archiving

  • Cassette surface texture optimizes scannability for archiving
TRUSTED_SUSTAINABILITY

Specifications

Pore Style Slots
Closure Included Yes
Quantity 750/Cs.
Closure Type Attached Lid, Front Hinge
PrintMate Compatible Yes
Color Beige
For Use With (Equipment) For Use With PrintMate™ Printers
Product Type Slotted Cassette
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.