missing translation for 'onlineSavingsMsg'
Learn More
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10269610

Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10269610
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10269610

Brand: Thermo Scientific Acros 424530250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-55-5
Molecular Formula C2H5NS
Molecular Weight (g/mol) 75.13
MDL Number MFCD00008070
InChI Key YUKQRDCYNOVPGJ-UHFFFAOYSA-N
Synonym thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amideShow MoreShow Less
PubChem CID 2723949
ChEBI CHEBI:32497
IUPAC Name ethanethioamide
SMILES CC(N)=S

Specifications

Melting Point 112.0°C to 114.0°C
Color White to Beige
Density 1.3700g/mL
Ignition Residue 0.05% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3CSNH2
Quantity 25 g
Beilstein 02, 232
Fieser 04,502
Merck Index 15, 9472
Specific Gravity 1.37
Solubility Information Solubility in water: 163g/L (25°C). Other solubilities: 26.4g/100g ethanol, sparingly soluble in ether, soluble in benzene
Formula Weight 75.13
Clarity (2% soln.) Passes test
Percent Purity 99+%
Grade ACS Reagent
Physical Form Crystalline Powder
Chemical Name or Material Thioacetamide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0452616 Certificate TypeCertificate of Analysis Date01/05/2024 Catalog Number424535000,424531000,424530250
Lot NumberA0444853 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number424535000,424531000,424530250
Lot NumberA0402854 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number424535000,424531000,424530250
Lot NumberA0431348 Certificate TypeCertificate of Analysis Date21/06/2022 Catalog Number424535000,424531000,424530250
Lot NumberA0426203 Certificate TypeCertificate of Analysis Date20/01/2022 Catalog Number424535000,424531000,424530250
compliance-icons compliance-icons
Product Identifier
  • Thioacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 1B
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P201-Obtain special instructions before use.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thioacetamide, 99+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.