missing translation for 'onlineSavingsMsg'
Learn More

Thioacetamide, ACS, 99% min

Product Code. 11357318
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11357318

Brand: Thermo Scientific Alfa Aesar 030638.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 62-55-5 | C2H5NS | 75.13 g/mol

Thioacetamide is used in qualitative inorganic analysis as an in-situ source for sulfide ions. It is used in analytical chemistry as a source of hydrogen sulfide. It is an additive in enantioselective reduction of beta-keto esters with immobilized bakers yeast. It is also used as a vulcanizing agent of polymer and a crosslinking agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-55-5
Molecular Formula C2H5NS
Molecular Weight (g/mol) 75.13
MDL Number MFCD00008070
InChI Key YUKQRDCYNOVPGJ-UHFFFAOYSA-N
Synonym thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
PubChem CID 2723949
ChEBI CHEBI:32497
IUPAC Name ethanethioamide
SMILES CC(N)=S

Specifications

Melting Point 112°C to 114°C
Quantity 50 g
Beilstein 506006
Merck Index 14,9319
Solubility Information Soluble in ethanol,benzene and petroleum ether. Slightly soluble in water and ether.
Formula Weight 75.13
Percent Purity 99%
Grade ACS
Physical Form Crystalline
Chemical Name or Material Thioacetamide
compliance-icons compliance-icons
Product Identifier
  • Thioacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Thioacetamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.