Learn More
Thioanisole, 99%, Thermo Scientific Chemicals
£23.60 - £358.00
Chemical Identifiers
CAS | 100-68-5 |
---|---|
Molecular Formula | C7H8S |
Molecular Weight (g/mol) | 124.21 |
MDL Number | MFCD00008559 |
InChI Key | HNKJADCVZUBCPG-UHFFFAOYSA-N |
Synonym | thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene |
PubChem CID | 7520 |
IUPAC Name | methylsulfanylbenzene |
SMILES | CSC1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10347070
|
Thermo Scientific Acros
138670250 |
25 g | Glass bottle |
£23.60
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10153570
|
Thermo Scientific Acros
138671000 |
100 g | Glass bottle |
£29.20
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10606152
|
Thermo Scientific Acros
138675000 |
500 g | Glass bottle |
£116.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10165823
|
Thermo Scientific Acros
138670025 |
2.5 kg | Glass bottle |
£358.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
100-68-5 | |
124.21 | |
HNKJADCVZUBCPG-UHFFFAOYSA-N | |
7520 | |
CSC1=CC=CC=C1 |
C7H8S | |
MFCD00008559 | |
thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene | |
methylsulfanylbenzene |
Specifications
100-68-5 | |
Brown-Yellow to Yellow | |
188.0°C | |
Authentic | |
C7H8S | |
1.5842 to 1.5862 | |
06, 297 | |
1.05 | |
Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents | |
CSC1=CC=CC=C1 | |
124.21 | |
124.21 | |
Liquid |
-15.0°C | |
1.0500g/mL | |
73°C | |
98.5% min. (GC) | |
C6H5SCH3 | |
MFCD00008559 | |
05,660; 06,582 | |
thioanisole, methyl phenyl sulfide, methylthio benzene, thioanisol, benzene, methylthio, phenylthiomethane, phenyl methyl sulfide, methylsulfanyl benzene, methylphenylsulfide, 1-thiaethyl benzene | |
HNKJADCVZUBCPG-UHFFFAOYSA-N | |
methylsulfanylbenzene | |
7520 | |
99% | |
Thioanisole, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-878-2
missing translation for 'rtecsNumber' : DA6200000
missing translation for 'tsca' : TSCA
- Thioanisole
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
Hazard Statement
- H317-May cause an allergic skin reaction.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only