Learn More
Thiobenzamide, 98%, Thermo Scientific Chemicals
£31.20 - £412.00
Chemical Identifiers
CAS | 2227-79-4 |
---|---|
Molecular Formula | C7H7NS |
Molecular Weight (g/mol) | 137.20 |
MDL Number | MFCD00008060 |
InChI Key | QIOZLISABUUKJY-UHFFFAOYSA-N |
Synonym | thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione |
PubChem CID | 683563 |
ChEBI | CHEBI:80418 |
IUPAC Name | benzenecarbothioamide |
SMILES | NC(=S)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11419843
|
Thermo Scientific Alfa Aesar
A12973.14 |
25 g |
£31.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429843
|
Thermo Scientific Alfa Aesar
A12973.22 |
100 g |
£101.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439843
|
Thermo Scientific Alfa Aesar
A12973.36 |
500 g |
£412.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thiobenzamide is useful for the preparation of amide and amidine adducts. It is also used to prepare 4-oxo-4H-chromene-3-carbothioic acid N-phenylamides. Further, it acts as a raw material in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiobenzamide is useful for the preparation of amide and amidine adducts. It is also used to prepare 4-oxo-4H-chromene-3-carbothioic acid N-phenylamides. Further, it acts as a raw material in organic synthesis.
Solubility
Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
2227-79-4 | |
137.20 | |
QIOZLISABUUKJY-UHFFFAOYSA-N | |
683563 | |
benzenecarbothioamide |
C7H7NS | |
MFCD00008060 | |
thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione | |
CHEBI:80418 | |
NC(=S)C1=CC=CC=C1 |
Specifications
2227-79-4 | |
120°C (248°F) | |
MFCD00008060 | |
606021 | |
Slightly soluble in water. | |
NC(=S)C1=CC=CC=C1 | |
137.20 | |
CHEBI:80418 | |
98% |
114°C to 118°C | |
C7H7NS | |
UN2811 | |
thiobenzamide, benzothioamide, benzamide, thio, benzothiamide, tiobenzamide, phenylthioamide, tiobenzamide italian, unii-8799vm9sxc, benzene-1-carbothioamide, aminophenylmethane-1-thione | |
QIOZLISABUUKJY-UHFFFAOYSA-N | |
benzenecarbothioamide | |
683563 | |
137.21 | |
Thiobenzamide |
Safety and Handling
GHS H Statement
H301
Toxic if swallowed.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H332-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 218-765-6
missing translation for 'rtecsNumber' : CV5860000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only