missing translation for 'onlineSavingsMsg'
Learn More

Thioflavine T, tech. 75%, Thermo Scientific Chemicals

Product Code. 15443709
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease

A dye used to visualize beta-amyloid plaques, like those associated with Alzheimer's Disease

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2390-54-7
Molecular Formula C17H19ClN2S
Molecular Weight (g/mol) 318.86
MDL Number MFCD00011944
InChI Key JADVWWSKYZXRGX-UHFFFAOYSA-M
Synonym Basic Yellow 1; C.I. 49005
PubChem CID 16953
ChEBI CHEBI:76023
IUPAC Name 4-(3,6-dimethyl-1,3-benzothiazol-3-ium-2-yl)-N,N-dimethylaniline;chloride
SMILES [Cl-].CN(C)C1=CC=C(C=C1)C1=[N+](C)C2=CC=C(C)C=C2S1

Specifications

Beilstein 3922452
Formula Weight 318.86
Percent Purity 75%
Grade Technical
Color Yellow
Quantity 5 g
Physical Form Powder
Chemical Name or Material Thioflavine T

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'