Learn More
Mercaptoacetic acid, 97+%, Thermo Scientific Chemicals
Air Senstitive
£29.35 - £211.05
Chemical Identifiers
CAS | 68-11-1 |
---|---|
Molecular Formula | C2H4O2S |
Molecular Weight (g/mol) | 92.112 |
MDL Number | MFCD00004876 |
InChI Key | CWERGRDVMFNCDR-UHFFFAOYSA-N |
Synonym | mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio |
PubChem CID | 1133 |
ChEBI | CHEBI:30065 |
IUPAC Name | 2-sulfanylacetic acid |
SMILES | C(C(=O)O)S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11494307
|
Thermo Scientific Alfa Aesar
B20391.22 |
100 g |
£29.35
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11404317
|
Thermo Scientific Alfa Aesar
B20391.36 |
500 g |
£46.83
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414317
|
Thermo Scientific Alfa Aesar
B20391.0E |
2500 g |
£211.05
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Reagent that protects tryptophan in amino acid analysis.Mercaptoacetic acid is used as a protecting agent for tryptophan in amino acid analysis and an acidity indicator. It finds application as an intermediate in the chemical reactions such as addition, elimination and cyclization. It acts as a precursor to ammonium thioglycolate, sodium thioglycolate and calcium thioglycolate. Its organotin derivatives are used as stabilizers for polyvinyl chloride (PVC). In organic synthesis, it acts as a nucleophile in thioglycolysis reactions and sulfur transfer agent for sulfonyl chloride synthesis. Further, it is used in leather processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsReagent that protects tryptophan in amino acid analysis.Mercaptoacetic acid is used as a protecting agent for tryptophan in amino acid analysis and an acidity indicator. It finds application as an intermediate in the chemical reactions such as addition, elimination and cyclization. It acts as a precursor to ammonium thioglycolate, sodium thioglycolate and calcium thioglycolate. Its organotin derivatives are used as stabilizers for polyvinyl chloride (PVC). In organic synthesis, it acts as a nucleophile in thioglycolysis reactions and sulfur transfer agent for sulfonyl chloride synthesis. Further, it is used in leather processing.
Solubility
Miscible with water, ethanol, ethers, ketones, esters, chlorinated hydrocarbons, benzene and aromatic hydrocarbons. Slightly miscible with chloroform.
Notes
Air sensitive. Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
68-11-1 | |
92.112 | |
CWERGRDVMFNCDR-UHFFFAOYSA-N | |
1133 | |
2-sulfanylacetic acid |
C2H4O2S | |
MFCD00004876 | |
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio | |
CHEBI:30065 | |
C(C(=O)O)S |
Specifications
68-11-1 | |
1.295 | |
119°C (246°F) | |
HSCH2CO2H | |
MFCD00004876 | |
506166 | |
14,9336 | |
Miscible with water,ethanol,ethers,ketones,esters,chlorinated hydrocarbons,benzene and aromatic hydrocarbons. Slightly miscible with chloroform. | |
C(C(=O)O)S | |
92.112 | |
CHEBI:30065 | |
≥97% | |
Mercaptoacetic acid, 98% |
°C to 12°C | |
220°C | |
C2H4O2S | |
1.5045 | |
UN1940 | |
Air Sensitive | |
mercaptoacetic acid, thioglycolic acid, 2-thioglycolic acid, acetic acid, mercapto, 2-mercaptoacetic acid, sulfanylacetic acid, thiovanic acid, mercaptoessigsaeure, thioglycollic acid, glycolic acid, thio | |
CWERGRDVMFNCDR-UHFFFAOYSA-N | |
2-sulfanylacetic acid | |
1133 | |
92.11 | |
Liquid |
Safety and Handling
GHS H Statement
H301-H311-H331-H314
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H301+H311+H331-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: THIOGLYCOLIC ACID
missing translation for 'einecsNumber' : 200-677-4
missing translation for 'rtecsNumber' : AI5950000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Mercaptoacetic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- Skin sensitiser Category 1
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Thioglycolic acid
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.