missing translation for 'onlineSavingsMsg'
Learn More

Thioisobutyramide, 95%, Thermo Scientific Chemicals

Código de producto. 15451959
Click to view available options
Quantity:
1 g
5 g
25 g
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 13515-65-6
Molecular Formula C4H9NS
Molecular Weight (g/mol) 103.183
MDL Number MFCD07369538
InChI Key NPCLRBQYESMUPD-UHFFFAOYSA-N
Synonym thioisobutyramide, isobutylthioamide, isobutyrthioamide, 2-methylpropaneothioamide, 2-methylpropane-thioamide, 2-methyl-propanethioamide, acmc-209by0, propanethioamide,2-methyl, ksc519m8d, 2-methyl propanethioamide
PubChem CID 10909507
IUPAC Name 2-methylpropanethioamide
SMILES CC(C)C(=S)N

Especificaciones

Melting Point 45°C to 48°C
Linear Formula (CH3)2CHC(=S)NH2
Quantity 25 g
Beilstein 1098427
Formula Weight 103.18
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Thioisobutyramide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'