missing translation for 'onlineSavingsMsg'
Learn More

Thionyl bromide, 97%, Thermo Scientific Chemicals

£30.85 - £63.92

Chemical Identifiers

CAS 507-16-4
Molecular Formula Br2OS
Molecular Weight (g/mol) 207.87
MDL Number MFCD00011448
InChI Key HFRXJVQOXRXOPP-UHFFFAOYSA-N
Synonym thionyl bromide, thionyl dibromide, unii-n6cu78b13t, thionylbromide, sulfur oxy dibromide, acmc-20ajin, pubchem2023, sobr2, thionyl bromide, purum at
PubChem CID 68176
IUPAC Name sulfurooyl dibromide
SMILES BrS(Br)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11352727
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L15648.14
25 g
£30.85
25g
Please to purchase this item. Need a web account? Register with us today!
11362727
View Documents Promotion Details
Thermo Scientific Alfa Aesar
L15648.22
100 g
£63.92
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thionyl bromide is used as a brominating agent in organic synthesis. It is used in the bromination of alfa-beta-unsaturated carbonyl compounds. It is also used to prepare alkyl bromides from alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thionyl bromide is used as a brominating agent in organic synthesis. It is used in the bromination of alfa-beta-unsaturated carbonyl compounds. It is also used to prepare alkyl bromides from alcohols.

Solubility
Miscible with benzene, toluene and ether.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with water, hydrobromide, acetone and alcohol.It hydrolyzes readily to release hydrogen bromide.
Specifications

Chemical Identifiers

507-16-4
207.87
HFRXJVQOXRXOPP-UHFFFAOYSA-N
68176
BrS(Br)=O
Br2OS
MFCD00011448
thionyl bromide, thionyl dibromide, unii-n6cu78b13t, thionylbromide, sulfur oxy dibromide, acmc-20ajin, pubchem2023, sobr2, thionyl bromide, purum at
sulfurooyl dibromide

Specifications

47°C to 49°C (20mmHg)
507-16-4
MFCD00011448
14,9347
Miscible with benzene,toluene and ether.
BrS(Br)=O
207.87
207.88
Acrid
Light and moisture sensitive
Thionyl bromide
-52°C
Br2OS
UN3264
thionyl bromide, thionyl dibromide, unii-n6cu78b13t, thionylbromide, sulfur oxy dibromide, acmc-20ajin, pubchem2023, sobr2, thionyl bromide, purum at
HFRXJVQOXRXOPP-UHFFFAOYSA-N
sulfurooyl dibromide
68176
97%
1.675
2.683
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H312-H314-H335

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: I

missing translation for 'einecsNumber' : 208-064-3

missing translation for 'tsca' : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only