missing translation for 'onlineSavingsMsg'
Learn More
Thionyl chloride, 99.5+%, Thermo Scientific Chemicals Product Code.: 10357490

Thionyl chloride, 99.5+%, Thermo Scientific Chemicals

Product Code. 10357490
500 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10357490

Brand: Thermo Scientific Acros 169495000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7719-09-7
MDL Number MFCD00011449
PubChem CID 24386
ChEBI CHEBI:29290

Specifications

Color Colorless through Yellow
Physical Form Liquid
Quantity 500 g
Assay Percent Range 99.5+%
Linear Formula SOCl2
Fieser 01,1158; 02,412; 03,290; 04,503; 05,663; 06,585; 07,366; 08,481; 09,465; 10,399; 12,204; 13,297
Formula Weight 118.97
Percent Purity ≥99.50%
Packaging Glass bottle
Density 1.6400g/mL
Chemical Name or Material Thionyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0440085 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog Number169490250,169495000,169490010,169491000,169490025
Lot NumberA0459941 Certificate TypeCertificate of Analysis Date05/02/2024 Catalog Number169490250,169495000,169490010,169491000,169490025
Lot NumberA0440085 Certificate TypeCertificate of Analysis Date19/04/2023 Catalog Number
Lot NumberA0434434 Certificate TypeCertificate of Analysis Date17/08/2022 Catalog Number169490250,169495000,169490010,169491000,169490025
Lot NumberA0428577 Certificate TypeCertificate of Analysis Date23/03/2022 Catalog Number169495000,169490250,169490010,169491000,169490025
compliance-icons compliance-icons
Product Identifier
  • Thionyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thionyl chloride, 99.5+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.