missing translation for 'onlineSavingsMsg'
Learn More

Thiophene-2,5-dicarboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 11308355
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.

Solubility
Very soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4282-31-9
Molecular Formula C6H4O4S
Molecular Weight (g/mol) 172.154
MDL Number MFCD00016896
InChI Key YCGAZNXXGKTASZ-UHFFFAOYSA-N
Synonym 2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid
PubChem CID 20273
IUPAC Name thiophene-2,5-dicarboxylic acid
SMILES C1=C(SC(=C1)C(=O)O)C(=O)O

Specifications

Melting Point 355°C to 360°C
Quantity 25 g
Beilstein 136781
Solubility Information Very soluble in water.
Formula Weight 172.16
Percent Purity 97%
Chemical Name or Material Thiophene-2,5-dicarboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'