Learn More
Thiophene-2,5-dicarboxylic acid, 97%, Thermo Scientific Chemicals
£16.10 - £144.00
Chemical Identifiers
CAS | 4282-31-9 |
---|---|
Molecular Formula | C6H4O4S |
Molecular Weight (g/mol) | 172.154 |
MDL Number | MFCD00016896 |
InChI Key | YCGAZNXXGKTASZ-UHFFFAOYSA-N |
Synonym | 2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid |
PubChem CID | 20273 |
IUPAC Name | thiophene-2,5-dicarboxylic acid |
SMILES | C1=C(SC(=C1)C(=O)O)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11388345
|
Thermo Scientific Alfa Aesar
L10293.03 |
1 g |
£16.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11398345
|
Thermo Scientific Alfa Aesar
L10293.06 |
5 g |
£45.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11308355
|
Thermo Scientific Alfa Aesar
L10293.14 |
25 g |
£144.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsThiophene-2,5-dicarboxylic acid may be used in the synthesis new cobalt(II) thiophenedicarboxylate coordination polymer and novel mesogenic thiophene based supramolecular liquid crystals.
Solubility
Very soluble in water.
Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Chemical Identifiers
4282-31-9 | |
172.154 | |
YCGAZNXXGKTASZ-UHFFFAOYSA-N | |
20273 | |
C1=C(SC(=C1)C(=O)O)C(=O)O |
C6H4O4S | |
MFCD00016896 | |
2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid | |
thiophene-2,5-dicarboxylic acid |
Specifications
4282-31-9 | |
C6H4O4S | |
136781 | |
Very soluble in water. | |
C1=C(SC(=C1)C(=O)O)C(=O)O | |
172.154 | |
172.16 | |
Thiophene-2,5-dicarboxylic acid |
355°C to 360°C | |
MFCD00016896 | |
2,5-thiophenedicarboxylic acid, 2,5-dicarboxythiophene, unii-5q8a2g9y8a, 2,5-thiophene dicarboxylic acid, pubchem5580, maybridge4_002021, acmc-1aj5p, ksc182i5d, 2,5-thiophene-dicarboxylic acid, thiophene-2.5-dicarboxylic acid | |
YCGAZNXXGKTASZ-UHFFFAOYSA-N | |
thiophene-2,5-dicarboxylic acid | |
20273 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 224-284-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only